Microtome, RM 2125 RTS, Leica
Supplier: Leica Biosystems MS
1492125RTS1
76001-006EA 21470.88 USD
76001-006 76404-240
Microtome, RM 2125 RTS, Leica
Microtomes and Microtome Assemblies Microtomes
Cut the high-quality sections that accurate diagnosis depends on. The Leica RM2125 RTS manual microtome has the essential features you need for safe, economic sectioning and optimized workflows.
  • Accuracy – precise feed mechanism means precise sections
  • Stability – PowerBase minimizes vibration during sectioning
  • Security – advanced knife holder and specimen clamp securely align cutting edge to block
When you’re cutting sections from patient tissue, you want every section to count. The Leica RM2125 RTS manual microtome utilizes the essential elements of proven Leica technology to deliver high-quality sectioning in an economical manual package.
Because you don’t want to compromise on safety, the RM2125 RTS manual microtome doesn’t either. By combining the essential elements of intuitive operation and advanced safety features, the RM2125 RTS manual microtome helps users work safely throughout each shift.
With no time to waste, efficient sectioning is critical for overall laboratory productivity. The Leica RM2125 RTS manual microtome focuses on essential features for highly productive sectioning.
Ordering information: For available options, configurations, and accessories (sold separately), contact your Avantor representative.
Delivery information: Basic instrument model – Basic instrument with X/Y specimen orientation and fast exchange system for specimen clamps, user selectable coarse feed wheel turn direction, retraction on/off function and two mechanical trim steps. The Leica RM2125 RTS rotary microtome has been designed for effortless manual sectioning via a counter-balanced, exceptionally smooth-running hand wheel. The object head with /-8° X/Y orientation is equipped with a fast specimen clamp exchange system for improved workflow. Storage space on top of the instrument housing provides room for sectioning tools and accessories.
Configuration 1 model – Basic instrument with X/Y specimen orientation and fast exchange system for specimen clamps, user selectable coarse feed wheel turn direction, retraction on/off function, two mechanical trim steps, blade holder for disposable blades and universal cassette clamp. The Leica RM2125 RTS rotary microtome has been designed for effortless manual sectioning via a counter-balanced, exceptionally smooth-running hand wheel. The object head with -/-8° X/Y orientation is equipped with a fast specimen clamp exchange system for improved workflow.