FreeZone® 12 L Console Freeze Dryers, Labconco®
Supplier: Labconco
701211010 701211015 701211030 701211040 701211050 701211070 701212010 701212015 701212030 701212040 701212050 701212070 701211110 701211115 701211130 701211140 701211150 701211170 701212110 701212115 701212130 701212140 701212150 701212170 701211210 701211215 701211230 701211240 701212250 701212270 711211010 711211015 711211030 711211040 711211050 711211070 711212010 711212015 711212030 711212040 711212050 711212070 711211110 711211115 711211130 711211140 711211150 711211170 711212110 711212115 711212130 711212140 711212150 711212170 711211210 711211215 711211230 711211240 711211250 711211270 711212210 711212215 711212230 711212240 711212250 711212270 701211250 701211270 701212210 701212215 701212230 701212240
76000-010EA 25425 USD
76000-010 76000-012 76000-014 76000-016 76000-018 76000-020 76000-022 76000-024 76000-026 76000-028 76000-030 76000-032 76000-034 76000-036 76000-038 76000-040 76000-042 76000-044 76000-046 76000-048 76000-050 76000-052 76000-054 76000-056 76000-058 76000-060 76000-062 76000-064 76000-066 76000-068 76000-070 76000-072 76000-074 76000-076 76000-078 76000-080 76000-082 76000-084 76000-086 76000-088 76000-090 76000-092 76000-094 76000-096 76000-098 76000-100 76000-102 76000-104 76000-106 76000-108 76000-110 76000-112 76000-114 76000-116 76000-118 76000-120 76000-122 76000-124 76000-126 76000-128 76000-130 76000-132 76000-134 76000-136 76000-548 76000-550 76000-660 76000-662 76000-664 76000-666 76000-668 76000-670
FreeZone® 12 L Console Freeze Dryers, Labconco®
Freeze Dryers
FreeZone 12 Liter Console Freeze Dryers are designed for lyophilizing moderate to large sample loads or numerous small container batches. Collectors are available to -50°C or -84°C.
  • Stay close to your samples as Lyo-Works™ operating system, provides mobile connectivity
  • Consistent, high quality results with flexible programming, and on board data collection
  • Challenging samples are easy with accessories designed specifically for laboratory samples
  • End-Zone™ End Point Detection option takes the guess work out of sample dryness
New Lyo-Works™ OS provides flexibility and convenience in an easy-to-use, touch screen display
The upright stainless steel collector coil holds 12 liters of ice before defrosting. Choice of a -50° C (for aqueous samples) or -84° C (for low eutectic point samples including acetonitrile) collector cooled by an environmentally-friendly HCFC/CFC-free refrigeration system. Internal coil maximizes vapor-trapping efficiency. Optional PTFE-coated collectors withstand corrosive compounds.
Lyo-Works OS provides a real time display of collector temperature and vacuum level on a 5"" full color touch screen. Flexible programming and control of optional drying accessories including End-Zone™ end point detection for flask freeze drying. One-touch data logging and display provides information to maximize sample quality and minimize run time. Customizable settings include language, time zone, temperature and vacuum. Alerts and feedback of current operating parameters can be sent automatically to user-entered email addresses.
FreeZone exclusive features protect your vacuum pump from costly damage. A vacuum break valve bleeds air into the system when power to the freeze dryer or vacuum pump is shut off for more than 5 minutes. A moisture sensor prevents refrigeration or vacuum start-up when moisture is detected in the drain line. Vacuum pump is automatically shut down when a leak is present in initial pull down or during run. Front viewing window makes monitoring vacuum pump oil hassle free.
Your samples are protected with remote notification and data recording. Features include hot gas defrost, side-mounted USB port and rear-mounted Ethernet connection. The console provides interior space for a vacuum pump and accommodates the FreeZone Stoppering Tray Dryer (pump and tray dryer sold separately).
Certifications: 60Hz models conform to UL Standard 61010-1 and CAN/CSA C22.2 No. 61010-1. 50 Hz models conform to CE Conformity marking.
Accessories information: Stainless steel lid have 14.0" dia., with neoprene gasket and 3" dia. hole for direct connection of drying accessories to collector.
Includes cap for 3" dia. attachment port. Acrylic lid have 14.0" dia., with neoprene gasket and 3" dia. hole for direct connection of drying accessories to collector. Includes cap for 3" dia. attachment port.
Ordering information: Vacuum pump with a displacement of at least 98 liters/minute, drying accessory and freeze dry glassware (if not bulk freeze drying) are required (not included).