Mouse Recombinant LIF (from E. coli)
Supplier: Peprotech
AF-250-02-5UG AF-250-02-25UG AF-250-02-100UG AF-250-02-250UG AF-250-02-500UG AF-250-02-1MG
75999-746EA 168.98 USD
75999-746 75999-748 75999-750 75999-752 75999-754 75999-756
Mouse Recombinant LIF (from E. coli)
Proteins and Peptides
LIF is a pleiotrophic factor produced by multiple cell types, including T cells, myelomonocytic lineages, fibroblasts, liver, heart and melanoma. LIF promotes long-term maintenance of embryonic stem cells by suppressing spontaneous differentiation. Other activities include the stimulation of acute phase protein synthesis by hepatocytes, stimulation of differentiation of cholinergic nerves, and suppression of adipogenesis by inhibiting the lipoprotein lipase in adipocytes. While human LIF is active on mouse cells and is widely used in the maintenance of murine ESC to prevent spontaneous differentiation, mouse LIF is not active on human cells due to its inability to bind to the human LIF receptor. Recombinant Murine LIF is a 19.9 kDa protein containing 180 amino acids residues, including three disulfide bonds.
  • Animal-free