Precision Mirco-Ceramic Cutter, Slice®
Supplier: SLICE, INC.
00116
75998-076CS 125.99 USD
75998-076
Precision Mirco-Ceramic Cutter, Slice®
Knives
The 00116 Precision cutter is a unique tool with a fixed micro-ceramic blade.
  • Blade type: micro-ceramic
  • Handle features non-slip finish
  • Ambidextrous design, reduces injuries, lowers costs
  • Finger-friendly® edge stays sharp 11x longer
  • Good Design® Award winner
  • Non-sparking, non-conductive blade
  • Chemically inert blade never rusts, oil, lubricant, and maintenance free
The cutter's tiny edge cuts thin materials and is well loved by artists and crafters. Its Zirconium oxide blade—much harder than steel—can handle thinner materials typically cut by craft knives but is safer to touch. The product weighs 21.2g.