Lysine-C™, Sequencing Grade, G-Biosciences
Supplier: G-Biosciences
786-14
75998-072EA 521.37 USD
75998-072
Lysine-C™, Sequencing Grade, G-Biosciences
Nucleic Acid Reagents Sanger Sequencing Reagents
SG-Lysine-C™ endopeptidase, from Lysobacter enzymogenes, is a serine protease highly specific in cleaving peptide bonds at the carboxy side of lysine. Highly purified preparations of SG-Lysine-C™ are chemically modified making the enzyme resistant to autolysis and stabilizing its enzymatic activity.
SG-Lysine-C™ is a modified Lysine-C with high specificity, free from other known endopeptidases, that is resistant to autolysis and degradation
SG-Lysine-C™ is supplied lyophilized in 5 μg vials. The enzyme is typically reconstituted to a concentration of 0.25 μg/mL. For fragmentation, the enzyme is added to the sample protein in a ratio of 1:100 to 1:20 (enzyme to protein, by weight) in a standard digestion buffer.
SG-Chymotrypsin™ is used for the digestion of proteins for sequence analysis and is suitable for sequencing applications. Use of SG-Lysine-C™ produces consistent activity levels from lot to lot for reproducible digestions.