VWR® DNA Molecular Weight Markers, 1 kb Ladder
Supplier: VWR
K181-1ML
75997-546EA 124.01 USD
75997-546
VWR® DNA Molecular Weight Markers, 1 kb Ladder
Electrophoresis Reagents Molecular Weight Markers
11 fragments ranging from 500 to 10,000 bp for easy band identification
  • Easy band identification
  • Available pre-mixed with loading dye
11 fragments ranging from 500 to 10,000 bp for easy band identification. Also available pre-mixed with loading dye or with EZ-Vision® One DNA dye.
Also available pre-mixed with loading dye or with EZ-Vision® One DNA dye.