Supermax Protege™ Stretch Nitrile Gloves
Supplier: SUPERMAX HEALTHCARE INC
93999
75997-484CS 250.37 USD
75997-484
Supermax Protege™ Stretch Nitrile Gloves
Gloves
Aurelia Protege Stretch Nitrile Powder Free Examination Gloves are made from 100% nitrile, ambidextrous, sky blue color and non-sterile.
  • 4.0 mil palm thickness
  • 100% latex free, rivals latex comfort and ergonomics
  • Fingertip-textured finish even grips extra-thin objects
  • Greater chemical resistance compared to the other materials
  • Ambidextrous, nonsterile, single use
Each batch of gloves is air-tested to detect pinholes. Aurelia gloves conform and are tested to the highest international standards.
Packaging: 100 gloves per box