Uriflex™ 24-HR Flexible Urine Collection Bag, Simport Scientific
Supplier: Simport Scientific
B351-4
75997-302CS 147.67 USD
75997-302
Uriflex™ 24-HR Flexible Urine Collection Bag, Simport Scientific
Bags Sample Bags
UriFlex™ 24-HR Flexible Collection Bag frees up inventory space with its flat bag design.
  • Bottom panel keeps bag in self-standing position
  • UV-resistant material perfect for analysis of light-sensitive substances
  • 50 mm wide screw closure with dripless valve
  • Large labeling area for easy patient identification
Made of co-extruded, laminated plastic film that will not rip, tear, or wrinkle even under heavy weight or stress. Strong seams virtually eliminate the possibility of leakage.
Certifications: CE marked.