Amplitude™ Knitted Wipes, Light weight, Contec®
Supplier: Contec
AMVL0001
75996-812CS 482.61 USD
75996-812
Amplitude™ Knitted Wipes, Light weight, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Amplitude knitted wipes are lightweight, 100% polyester interlock knit wipes that feature a soft, textured surface for effective cleaning without scratching.
  • Wipes are laser cut, which seals the fibers at the edges
  • Wipes are made with fabric knitted from filament polyester
They have good sorbency with solvents and are extremely low in particles and fibers. Wipes are laundered and packaged in an ISO Class 4 cleanroom to further reduce contamination.