Fluoride Test Papers, CTL Scientific Supply
Supplier: Macherey-Nagel
F-100
75996-722BX 1865.92 USD
75996-722
Fluoride Test Papers, CTL Scientific Supply
Test Paper Indicator Paper
Used for the determination of fluoride ions and gaseous hydrogen fluoride (H2F2).
  • Pinkish-red paper turns yellowish-white in the presence of fluorides
  • Sensitivity limit 20 mg/L fluoride
Substantial amounts of chlorates, bromates and sulphates cause white discolorations of the paper. These interferences are readily circumvented.