DURAN® TILT Media Bottle System, DWK Life Sceinces
Supplier: DWK Life Sciences (KIMBLE)
218914459
75996-374CS 171.44 USD
75996-374
DURAN® TILT Media Bottle System, DWK Life Sceinces
Bottles Cell Culture Bottles
The DURAN® TILT bottle functions in two positions; upright for filter sterilization or storage, and tilted at a 45° angle for pipetting. Take a look at cell culture media from a new angle and discover a bottle system that eases handling and turns usability into security.
A full range of accessories for the DURAN® TILT media bottle system are available.
DURAN® TILT GL 56 self-adhesive cap labels allow careful labelling and prevent mix-ups and mistakes. The DURAN® TILT Protective Light Shield is a white silicone sleeve that covers 94% of the bottle surface, blocking damaging ultraviolet (UV) light, protecting the glass surface from damage, and facilitating safer handling. The silicone sleeve provides a much better grip than glass; especially when the bottle has been sprayed with 70% alcohol for surface disinfection. DURAN® GL 56 Bottle Tags can be used on their own for color identification or to secure the protective Light Shield around the bottle (orange, yellow, blue, and purple tags included). DURAN® Bottle Top Vacuum Filtration Units are designed for sterilization or clarification of aqueous cell culture media. Supplied as a filter funnel-only unit for use with the DURAN® TILT bottle (with GL 45 thread adaptor) or 45 mm media bottles. The reusable adaptor (GL 45 external/GL 56 internal) allows the use of the DURAN® TILT bottle with 45 mm filtration units for the filter sterilization of cell culture media. Manufactured from inert PTFE, they can be autoclaved and depyrogenised at 300 °C.