Guthrie Double Hook
Supplier: Sklar
98-821
75996-350EA 23.8 USD
75996-350
Guthrie Double Hook
Retractors
Merit™ Guthrie Hook is a retractor used in delicate procedures like plastic surgery to reduce trauma.
  • Reusable
  • Nonsterile
The two prongs are delicate and sharp with a 1.5 mm width between them. The overall length is 5 inches.