YMC-Triart C8 Hybrid Silica HPLC Columns, C8, 3 µm, YMC America
Supplier: YMC America,Inc
TO12S03-0501WT TO12S03-0502WT TO12S03-0503WT TO12S03-0506WT TO12S03-0546WT TO12S03-1001WT TO12S03-1002WT TO12S03-1003WT TO12S03-1006WT TO12S03-1046WT TO12S03-1502WT TO12S03-1503WT TO12S03-1546WT TO12S03-H501WT TO12S03-H502WT TO12S03-H546WT TO12S03-L502WT TO12S03-L506WT TO12S03-L546WT TO12S03-0202WT TO12S03-0302WT TO12S03-2502WT TO12S03-2546WT TO12S03-05Q1PTP TO12S03-10Q1PTP TO12S03-15Q1PTP TO12S03-0546PTP TO12S03-1046PTP TO12S03-1546PTP TO12S03-L503WT
89217-042EA 1037.4 USD
89217-042 89217-044 89217-046 89217-048 89217-050 89217-052 89217-054 89217-056 89217-058 89217-060 89217-062 89217-064 89217-066 89217-068 89217-070 89217-076 89217-078 89217-080 89217-082 89217-084 89217-086 89217-088 10842-224 10842-232 10842-234 10842-236 75995-148 75995-150 75995-152 75995-154 75995-156 75995-158 76013-932
YMC-Triart C8 Hybrid Silica HPLC Columns, C8, 3 µm, YMC America
Chromatography Columns
YMC-Triart C8, 3µm is a novel organic/inorganic hybrid silica particle derivatized with C8 (L7 USP designation).
  • Suitable for HPLC
  • Chemical and mechanical durability of the particles contributes to long column lifetime
  • Proprietary end capping makes for excellent peak shapes of all types of compounds
  • Rigorous manufacturing controls result in legendary YMC column-to-column and lot-to-lot reproducibility
  • More retentive than other hybrid materials, potentially simplifying the separation of 'difficult' peaks
  • YMC-Triart C8 is available in packed columns of various dimensions
The core material for all of the YMC-Triart products, the particle, is inspired by YMC’s flow chemistry technology. Inherent in YMC’s process is a high level of temperature and particle formation control. As a result, Triart particles are consistent, reproducible, and scalable across many particle sizes. The hybrid Triart particles produce a material that is highly resilient and robust when used at extreme pH and elevated temperature. In addition, these particles are very stable under mechanical stress such as high pressure. YMC-Triart C8 can be used at temperatures up to 70 °C at pH 1 to 7, and 50 °C from pH 7 to 12.
C8 is an alternative to the more widely-used C18 chemistry, and is of lower hydrophobicity than C18. C8’s lesser retention may help shorten total analysis times in separations of compounds with large differences in hydrophobicity without requiring gradient elution.