Electrothermal CMUA Series Stirring, Heating Mantles, Cole-Parmer
Supplier: Antyila Scientific
36013-13
75994-960EA 1806.75 USD
75994-960 75994-962 75994-964
Electrothermal CMUA Series Stirring, Heating Mantles, Cole-Parmer
Heaters Heating Mantles
Magnetic stirrer and heating mantle in one convenient unit.
  • Flexible coiled heating element wraps the heater around round-bottom flask to optimize heat transfer
  • 'Cool to the touch' exterior provides safe handling
These stirring, heating mantles combine the functionality of a heating mantle with a magnetic stirrer providing even heating and mixing. Element temperatures can reach up to 825 °F (450 °C) yet the exterior remains cool to the touch. For maximum safety, the flask does not come in direct contact with the heating element.
Built-in energy regulator and safety earth (ground) screen offer added safety protection. Reach stirring speeds up to 2000 rpm. Simply use the auto-recapture switch to reactivate stirring if stirring is interrupted and stirring motion is lost. Two colored lights by each control dial indicate 'power on' and 'heat on'. Powder-coated aluminum housing makes it easy to clean.