Cole-Parmer® RS-250 Series Reaction Stations, Antylia Scientific
Supplier: Antyila Scientific
99968-13
75994-938EA 13305.21 USD
75994-938 75994-940 75994-942
Cole-Parmer® RS-250 Series Reaction Stations, Antylia Scientific
Stirrers Magnetic Hotplate Stirrers
Run multiple reactions in a single station, saving time and money.
  • Excellent temperature uniformity across the block
  • Soft-start feature minimizes splashing
Run these reaction stations at the same temperature and stirring rates at several positions simultaneously. They are great for a wide range of applications from simple synthesis to process optimization. High-performance magnetic stirrers ensure maximum coupling between the stir bar in the sample and motor.
Easily set and view temperature and stir rate on the digital display. Use the RS232 or RS485 connections to interface with a robotic workstation or a computer. The well-insulated stations stay cool to the touch for safe handling. PTFE coating on vessel block provides protection from chemical spills.