Indoxyl Acetate Disks, rapid test for Campylobacter, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z111
75993-556EA 105.67 USD
75993-556
Indoxyl Acetate Disks, rapid test for Campylobacter, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
For the identification and differentiation of Campylobacter, Wolinella and Helicobacter.
Look for a blue to blue-green color within only 30 minutes.