GlabrataQuick Kit, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z298
75993-498EA 450.59 USD
75993-498
GlabrataQuick Kit, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
GlabrataQuick™, for the identification of Candida glabrata, 24 tests
  • Test within one to two hours
  • Greater accuracy
  • Easy color read-out
GlabrataQuick™, contains two 96 well trays and 24 Inoculation Buffer tubes with 1ml fill to complete a total of 24 tests. GlabrataQuick™ will accurately identify Candida glabrata within one to two hours. It tests for three carbohydrates for greater accuracy; trehalose, sucrose, and maltose. Easy and distinct color read-out. Carbohydrates turn from blue to yellow.