HardyDisks™ Piperacillin/Tazobactam, TZP-110, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z8915
75993-460EA 83.51 USD
75993-460
HardyDisks™ Piperacillin/Tazobactam, TZP-110, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
  • Impregnated disks
  • Spring loaded dispenser
  • Smooth and reliable dispensing
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.