HardyDisks™ Cefotetan, CTT-30, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z8235
75993-458EA 81.19 USD
75993-458
HardyDisks™ Cefotetan, CTT-30, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
  • Impregnated disks
  • Spring loaded dispenser
  • Smooth and reliable dispensing
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.