HardyDisks™ Ampicillin/Sulbactam, SAM-20, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z8045
75993-456EA 87.7 USD
75993-456
HardyDisks™ Ampicillin/Sulbactam, SAM-20, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
  • Impregnated disks
  • Spring loaded dispenser
  • Smooth and reliable dispensing
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.