PL-E Portable Precision Balances, METTLER TOLEDO®
Supplier: Mettler Toledo
30113823
75990-508EA 1435 USD
75990-508 75990-510 75990-580
PL-E Portable Precision Balances, METTLER TOLEDO®
Balances and Scales
METTLER TOLEDO PL-E Portable Precision balances offer reliable results for on-the-go weighing. These balance are compact and light–ideal when accurate results are needed in tight spaces.
  • Capacities from 620 g to 6200 g
  • Readabilities from 0.01 g
  • Tough and lightweight design
  • Compact and portable
  • Stackable cover
Vibration resistant design provides good performance on temporary work surfaces and in the field.
Every PL-E model can be DC or battery operated, making the balance ideal for work whenever a power supply is hard to find.
Stack and pack up to 5 balances for quick, easy storage – effectively protecting them against dust and dirt.
Certifications: UL listed and CSA certified.