OsmoPRO® Multi-Sample Micro-Osmometer, Advanced Instruments
Supplier: Advanced Instruments
OSMOPRO
75990-506EA 100962.28 USD
75990-506
OsmoPRO® Multi-Sample Micro-Osmometer, Advanced Instruments
Clinical Diagnostic Supplies Clinical Analysers
This osmometer features a convenient carousel design for easy loading of up to 20 samples and requires only a small 20 µL sample volume
  • Large 7" touchscreen display
  • Fast and accurate osmolality test results
  • Easy printout and archiving of test results
  • True 3 point calibration
  • Multi-language operation, on board statistical analysis and an internal memory of the last 1000 samples
It is ideally suited for mid- to high-volume laboratories that prefer to automate osmolality batch testing while improving laboratory efficiency and throughput.