VWR® Pouring Ring for Bottles
Supplier: VWR International
275.205.02W
75983-266CS 434.42 USD
75983-266
VWR® Pouring Ring for Bottles
Closures Bottle Closures
Made from polyethylene.
  • Autoclavable up to 140 °C
  • White color
For GL45 storage bottle.