VWR® 20 mm Crimp Seal Headspace Vials
Supplier: VWR International
75983-244 75983-246 75983-248 75983-250 75983-252 75983-254 75983-256
75983-244PK 67.88 USD
75983-244 75983-246 75983-248 75983-250 75983-252 75983-254 75983-256
VWR® 20 mm Crimp Seal Headspace Vials
Vials
Clear 20 ml headspace vials designed to fit most headspace autosamplers.
  • Round bottom vials fit PerkinElmer®, Tekmar®, and Varian® headspace autosamplers
  • Flat bottom vials fit Agilent® and Carlo Erba autosamplers
Ordering information: 20 mm crimp seals have to be ordered separately.