Heating Pads, AIMS™
Supplier: Aims
GGP1
75983-092EA 48.63 USD
75983-092
Heating Pads, AIMS™
Heaters Hot Plates
Gel heating pad to be used with standard thermal plate and can be reactivated.
  • Maintains temperature around 33 to 34 °C