Peggy Repetitive Pipettor, Watson Bio Lab
Supplier: Watson Bio Lab
PG-01
75982-726EA 452.71 USD
75982-726
Peggy Repetitive Pipettor, Watson Bio Lab
Pipettes Pipette Controllers
This handy manual repetitive pipettor doesn't need to recharge or use a battery. To be used with disposable pipettes (5 / 10 ml).
  • Flexible nozzle angle
  • Simple maintenance
  • Lightweight and easy to carry
  • Aspiration can be set in steps of 1 ml for setting range 1 - 13 ml
  • Dispensing can be set in steps of 0.5 ml for setting range 0.5 - 5 ml
The pipettor can aspirate all at once and discharge necessary volume repetitively. The nozzle adjusts angles every 10° up to 50°, and spare parts are easy to remove and set.