VWR® Chemical Splash Goggles
Supplier: VWR International
G-03
75982-628CS 2253.78 USD
75982-628
VWR® Chemical Splash Goggles
Goggles
Wide vision goggles provide high levels of comfort and maximum splash protection.
  • Wraparound modern style provides excellent panoramic vision
  • Fits most facial contours
  • Comfortably fits over most prescription glasses
  • Indirect ventilation
  • Provides 99.9% UV protection
These high-quality safety goggles feature a modern, wraparound style with fogless, distortion-free panoramic lenses. They are equipped with indirect venting for liquid splash protection and internal relief slots to reduce interference with prescription glasses. The wide elastic strap with adjustable buckle attaches to the frame with a swivel clip for a comfortable fit. The nose area is split for greater comfort and the wide facial flange provides a secure seal to almost every face.
Certifications: Meet ANSI Z87+ 1-2015 standards.