PIG® Grippy® Plunger Mat, New Pig
Supplier: New Pig
GRP7005-BK
75982-604BX 19.64 USD
75982-604
PIG® Grippy® Plunger Mat, New Pig
Floor Mats Tacky Mats
Control messes from toilet plungers and brushes with adhesive-backed PIG® Grippy® Plunger Mats for a more professional restroom environment.
  • Place underneath toilet plungers, brushes, soap dispensers, and other small-footprint areas in the restroom to control drips and eliminate puddles
  • Proprietary adhesive formulation applied across entire backing of mats ensures they stay in place with no shifting, curling, or flipping up
  • Permanently infused with antimicrobial technology to prevent the growth of odor-causing bacteria and mold
  • Top surface is engineered to
More Product Information