SureSTART™ WebSeal™ 96-Well Mid-well Plates, Square U-Bottom
Supplier: Thermo Scientific
60180-P155
75982-330PK 775.48 USD
75982-330
SureSTART™ WebSeal™ 96-Well Mid-well Plates, Square U-Bottom
Microplates Solid Supported Liquid/Liquid Extraction Plates (SLE)
SureSTART™ Level 1 WebSeal™ 96-Well Square U-Bottom Mid-well plates are designed for everyday chromatography analysis.
  • Ideal for non-regulated labs that use water and polar solvents
  • Commodity plates, seals, tapes
  • GC tested
  • Suitable for HPLC/UHPLC applications
  • Suitable for biopharmaceutical applications
  • Suitable for QA/QC applications using core detectors
  • Ideal for verifying compound presence and quantification using detection techniques such as UV, DAD/PDA, RI, Fluorescence, and ELSD
  • SureSTART WebSeal well plates are dimensionally verified
  • Non sterile and non coated
  • 8×12 array
They provide an affordable choice for use with most HPLC/UHPLC instrument types, and are ideal for non-regulated laboratories using water, water and polar solvents and polar solvents. These 96-well plates are specification certified to ensure autosampler compatible physical dimensions and conform to ANSI-SLAS 4-2004 standards.
Certifications: GC/MS Tested.