BioClean™ Legacy Latex Gloves, Ansell
Supplier: Ansell Healthcare
BLA204-XL BLA205-XXL BLA200-XS BLA201-S BLA202-M BLA203-L
75982-142CS 712.17 USD
75982-142 75982-144 75982-182 75982-298 75982-300 75982-302
BioClean™ Legacy Latex Gloves, Ansell
Gloves
Gloves provide wearer comfort with flexibility, they can be double-donned and feature a textured surface for enhanced grip, and a beaded cuff for strength and stability on the arm. These ambidextrous cleanroom gloves with beaded cuffs can be used in medical as well as in laboratory environments
  • Protein level less than 50 µg/g
  • Textured finish for improved grip
  • Latex provides exceptional flexibility and comfort
  • Beaded cuff provides easier donning and stability on the arm
  • Each size carries a unique barcode on the carton (ITF-14), which allows automatic data entry into stock control systems
  • Suitable for double-donning
Suitable for use in Class 10, ISO 4 or M2.5 cleanrooms. Suitable for contact with dry, alcoholic and acidic foodstuffs.
Certifications: Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425 Module D certifications. Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Regulation (EU)2016/425. Processed in an NEBB certified ISO Class 4 environment. Complies with the requirements of European Glove Standards EN 420:2003 +A1:2009, EN 374-1:2016, EN 374-5:2016 and EN 421:2010. Physical properties comply with European Medical Glove Standard EN 455-2:2015.Regulation (EC)1935/2004 and (EC) 2023/2006.
Packaging: 100 pieces per sealed inner PE bag, one sealed inner PE bag per sealed outer PE bag, 10 outer PE bags per carton liner, one carton liner per carton (1000 pieces).