BioClean-D™ Sterile Cleanroom Lab Coats, Ansell
Supplier: Ansell Healthcare
S-BDLC-L S-BDLC-M S-BDLC-S S-BDLC-XL S-BDLC-XXL
75982-286CS 586.46 USD
75982-286 75982-288 75982-290 75982-292 75982-294
BioClean-D™ Sterile Cleanroom Lab Coats, Ansell
Disposable Garments
BioClean-D™ sterile single-use lab coats are manufactured from antistatic lightweight CleanTough™ material.
  • Press stud fastening
  • Open cuffs
  • Three deep pockets
  • rear vent
  • Excellent ESD properties
  • Latex-free
Lightweight CleanTough™ material is low linting and provides reliable anti-static protection. Garments feature bound seams with single needle stitching. These sterile lab coats are gamma irradiated to an Sterility Assurance Level (SAL) of 10⁻⁶.
Certifications: ASTM F739-12, EN ISO 13034:2005+A1:2010, EN 6530.