BioClean-D™ Sterile Coveralls with Collar, Ansell
Supplier: Ansell Healthcare
S-BDCCT-L S-BDCCT-M S-BDCCT-S S-BDCCT-XL S-BDCCT-XXL S-BDCCT-XXXL 850455
75982-188CS 200.14 USD
75982-188 75982-190 75982-192 75982-194 75982-196 75982-198 75982-200
BioClean-D™ Sterile Coveralls with Collar, Ansell
Disposable Garments
The BioClean-D™ coverall with collar.
  • Lightweight CleanTough™ material
  • Thumb loops to ensure a secure hold
  • Front zip with sealable cover and protective flap
  • Elasticated back, cuffs, and ankles
  • Excellent ESD properties
  • Latex-free
The anti-static, low-linting CleanTough™ material provides comfort and protection from a range of chemicals. Lightweight yet durable garments feature bound seams with single needle stitching. ISO Class 4 cleanroom compatible. These sterile garments are gamma irradiated to an Sterility Assurance Level (SAL) of 10⁻⁶.
Certifications: ASTM F739, EN ISO 13688:2013, EN 1 ASTM F739, EN ISO 13688:2013, EN 14325, Sterilized in accordance with ISO 11137-1:2006.