Slide Warmer with Anodized Black Surface, Azer Scientific
Supplier: VWR International
75981-544
75981-544EA 589.42 USD
75981-544
Slide Warmer with Anodized Black Surface, Azer Scientific
Slide Warmers/Dryers
This simple slide warmer is easy to operate and is ideal for smaller throughput histology labs.
  • 23 slide capacity
  • Contains 1 anodized black surface for easy visualization and even heating
  • Features a digital LED display
This slide warmer is ideal for use in small to medium throughput histology labs. Its anodized black surface offers even heating for slides mounted with tissue sections. Simply turn switch "On" to begin drying.