Slide Warmer with Customizable Tiers, Azer Scientific
Supplier: VWR International
ES4573
75981-542EA 987.22 USD
75981-542
Slide Warmer with Customizable Tiers, Azer Scientific
Slide Warmers/Dryers
This multi-tiered slide warmer allows users to easily add and remove slides without interfering with other slides on the warmer.
  • 40 slide capacity
  • Contains 4 tiers
This slide warmer offers 4 tiers for drying up to 40 microscope slides at a time. Its design allows users to easily add or remove slides without interfering with others.