Slide Warmer with LED Display, Azer Scientific
Supplier: VWR International
75981-540
75981-540EA 765.98 USD
75981-540
Slide Warmer with LED Display, Azer Scientific
Slide Warmers/Dryers
This sleek, modernized slide warmer is ideal for use in histology labs.
  • 66 slide capacity
  • Features an LED temperature display
  • Anodized black surface provides helpful contrast
This slide warmer is ideal for use in the histology laboratory for slides mounted with tissue sections. The thermal heater ensures that heat is distributed evenly across the surface.