Anti-Rabies Mouse Monoclonal Antibody [clone: Rab-50]

Supplier: Biotium

BNUM0943-50
75975-448EA 423.94 USD
75975-448
Anti-Rabies Mouse Monoclonal Antibody [clone: Rab-50]
Antibodies
This antibody recognizes viral glycoprotein of rabies virus strains SAD-Vnukovo and Pitman-Moore. This MAb is useful in detecting rabies virus by ELISA and Western. It is capable of neutralizing rabies virus. Rabies virus (Neurotropic virus) is a member of the Rhabdoviridae family. Rabies is a single stranded, neurotropic, negative sense RNA virus which encodes 5 proteins: a glycoprotein, a nucleoprotein, and three others. The mature virus has a bullet shape, a protein coat, and a lipid envelope. The outer surface of the virus is covered with thumb like glycoprotein projections 5-10 nm long and 3 nm in diameter. The virus averages approximately 780 nm in length. Lipid solvents destroy virus infectivity. Rabies virus is a very successful virus, with a very wide range of hosts. It causes an acute, central nervous system infection, characterized by CNS irritation, followed by paralysis and death. Approximately 50,000 human deaths each year are caused by rabies.

Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: Clone Rab-50
Conjugation: Unconjugated, BSA-and azide-free
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Rabies
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR