BioClean-D™ Non sterile Coveralls with Hood, Ansell
Supplier: Ansell Healthcare
BDCHT-L BDCHT-M BDCHT-S BDCHT-XL BDCHT-XS BDCHT-XXL BDCHT-XXXL BDCHT-XXXXL
75972-550CS 363.26 USD
75972-550 75972-552 75972-554 75972-556 75972-558 75972-560 75972-562 75972-656
BioClean-D™ Non sterile Coveralls with Hood, Ansell
Disposable Garments
The BioClean-D™ coverall with hood is a disposable garment featuring a front zip with protective flap, elasticated hood, back, cuffs and ankles, and thumb loops to ensure a secure hold.
  • Lightweight CleanTough™ material
  • Thumb loops to ensure a secure hold
  • Front zip with sealable cover
  • Elasticated hood, back, cuffs, and ankles
  • Excellent ESD properties
  • Latex-free
The anti-static, low-linting CleanTough™ material provides comfort and protection from a range of chemicals. Lightweight yet durable garments feature bound seams with single needle stitching. ISO Class 4 cleanroom compatible.
Certifications: ASTM F739, EN ISO 13688:2013, EN 1 ASTM F739, EN ISO 13688:2013, EN 14325.