SUMO2
Supplier: Rockland Immunochemical
009-001-T67
75935-530EA 210 USD
75935-530 75935-532
SUMO2
Proteins and Peptides
SUMO2 control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
SUMO2 is a member of the SUMO (small ubiquitin-like modifier) protein family which functions in a manner similar to ubiquitin. SUMO2 is bound to target proteins as part of a post-translational modification system and modifies numerous cellular proteins and affect their metabolism and function. SUMO2 is also involved in a variety of cellular processes, such as nuclear transport, transcriptional regulation, apoptosis, and protein stability (1). SUMO2 is predominantly detected on nuclear bodies (2). SUMO2 Protein is ideal for investigators involved in Signaling Proteins, Ubiquitin Proteins, Apoptosis/Autophagy, Cell Cycle, and NfkB Pathway research.
SUMO2 Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. SUMO2 Protein is suitable for use in Western Blot. Expect a band approximately ~17.5kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.