3,3',5,5'-Tetramethylbenzidine (TMB)

Supplier: Rockland Immunochemical

Synonyms: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine

TMBE-20L TMBE-8000
75935-450EA 4312.84 USD
75935-450 75935-452
3,3',5,5'-Tetramethylbenzidine (TMB)
3,3',5,5'-Tetramethylbenzidine (TMB)

TMB ELISA Peroxidase Substrate (3, 3', 5, 5' - Tetramethylbenzidine) is a chromogenic substrate used to visualize antibody reactivity in ELISA experiments. In the presence of peroxidases, TMB can act as an electron donor for the conversion of peroxides to water, changing the color of solution to a blue color equivalent to the degree of reactivity. This reaction can be halted with acid to change the TMB to yellow. TMB ELISA Peroxidase Substrate is ideal for investigators in Microbiology, Immunology, and Cancer research.

Formula: C₁₆H₂₀N₂
MW: 240.35 g/mol
Boiling Pt: 402
Storage Temperature: Ambient
MDL Number: MFCD00007748
CAS Number: 54827-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR