Anti-SETD8 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
100-401-V82
75935-246EA 520.3 USD
75935-246
Anti-SETD8 Rabbit Polyclonal Antibody
Antibodies
SETD8 is a histone methyl transferase that specifically monomethylates 'Lys-20' of histone H4. This histone modification represents a specific tag for epigenetic transcriptional repression and is enriched during mitosis. SETD8 may also play a role in maintaining silet chromatin by preventing the acetylation of the H4 tail. Anti-SETD8 Antibody is ideal for research in Gene Expression, Epigenetics and Chromatin Remodeling.

Anti-SETD8 Antibody is suitable for Western Blots. Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 40 kDa in the appropriate cell lysate or extract.

Type: Primary
Antigen: SETD8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR