Rat Tf
Supplier: Rockland Immunochemical
012-0134
75935-202EA 283.13 USD
75935-202
Rat Tf
Proteins and Peptides
Rat Transferrin
  • control or standard reagent
  • SDS, Western Blotting, and ELISA experiments
Rat Transferrin is a blood plasma glycoprotein which binds iron and regulates the amount of available iron in circulation. Rat Transferrin has a high affinity for iron, but this binding is reversible. Rat Transferrin has a molecular weight of approximately 80kDa. Rat Transferrin also plays a role in the immune system, creating environments low in iron for which many pathogenic bacteria are unable to thrive.
Rat Transferrin is a suitable protein for use as a control reagent in both Western Blotting and ELISA experiments. For blotting, the expected molecular weight of Rat Transferrin is approximately 80kDa.