Anti-Rptor Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-DW6
75935-140EA 635.22 USD
75935-140
Anti-Rptor Rabbit Polyclonal Antibody
Antibodies
The mammalian Target of Rapamycin (TOR, also known as mTOR) is an evolutionarily conserved serine/threonine kinase that regulates cell growth and cell cycle through its ability to integrate signals from nutrient levels and growth factors. Rapamycin inhibits TOR activity resulting in reduced cell growth and reduced rates of cell cycle and cell proliferation. Raptor (regulatory associated protein of TOR) is a TOR-binding protein essential for TOR signaling in vivo. It acts as a TOR scaffold protein whose binding by TOR substrates is necessary for effective TOR-catalyzed phosphorylation. These substrates include the ribosomal protein S6 kinase (RP S6K) and the eukaryotic initiation factor 4E binding protein 4EBP1, proteins necessary for cell growth and proliferation and responsive to nutrient and mitogen levels. Raptor binds these proteins through a common 5 amino acid TOR-signaling (TOS) motif; mutation of this motif prevents the TOR-dependent phosphorylation of these proteins.

Anti-Raptor Antibody has been tested for use in ELISA, Western Blotting, Immunocytochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 149 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: Rptor
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR