Rabbit 'B' (Non-Pooled) Complement (Fresh Frozen), Rockland Immunochemicals
Supplier: Rockland Immunochemical
C308
75935-070EA 763.73 USD
75935-070
Rabbit 'B' (Non-Pooled) Complement (Fresh Frozen), Rockland Immunochemicals
Sera (Serum)
Complement Fresh Frozen
  • prepared by removal of fibrinogen through clot formation under proper temperature conditions
  • Complement is preservative FREE
Special processing techniques are used to yield products with high complement activity and low background cytotoxicity. Rabbit "B" Complement is suitable for CFT and SRH.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal
Immunoelectrophoresis: normal