Anti-PUUVsSgp1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-DV6
75935-062EA 635.22 USD
75935-062
Anti-PUUVsSgp1 Rabbit Polyclonal Antibody
Antibodies
Puumala virus (PUUV) is a rodent-borne hantavirus of the family Bunyaviridae, an enveloped, negative-sense RNA viruses with a tripartite genome that can cause hantavirus pulmonary syndrome (HPS) and is highly homologous to the protype hantavirus Hantaan virus (1). Like other hantaviruses, the PUUV glycoprotein is synthesized as a precursor that is posttranslationally processed into two glycoproteins G1 (Gn) and G2 (Gc). These glycoproteins interact with the PUUV nucleocapsid (NP) protein through their cytoplasmic tail, and this association has been suggested to be crucial to the binding of the ribonucleoprotein of the PUUV and the assembly of the virus particle (2).

Anti-Puumala Virus Nucleocapsid Antibody has been tested for use in ELISA. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 49 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: PUUVsSgp1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR