Anti-LMW E.Coli Host Cell Proteins Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-401-J06S
75934-870EA 151.49 USD
75934-870
Anti-LMW E.Coli Host Cell Proteins Rabbit Polyclonal Antibody
Antibodies
Anti-LMW Host Cell Proteins Antibody recognizes low molecular weight protein fractions from E.coli. This antibody has been anlyzed by 2D electrophoresis western blot to assure highest possible coverage of E.coli HCP. Host Cell Proteins are process-related impurities derived from a host cell expression system that may be present in trace amounts in a final drug substance. HCP Antibodies are used to detect impurities in biologically-based therapeutics. Detecting impurities often unseen by 1-D electrophoresis, a 2D electrophoresis western blot analysis delivers more complete insight into the immunocoverage of protein components of the host cell lysate.

Anti-LMW Host Cell Proteins Antibody detects over 270 E.coli LMW proteins and is suitable for western blotting and ELISA.

Type: Primary
Antigen: LMW E.Coli Host Cell Proteins
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR