Anti-LAMP1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-CH1
75934-818EA 635.22 USD
75934-818
Anti-LAMP1 Rabbit Polyclonal Antibody
Antibodies
Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components and is negatively regulated by TOR (Target of rapamycin). A protein recently found to be involved in autophagy, LAMP-2, is a highly glycosylated protein associated with the lysosome. LAMP-1 shares much homology to LAMP-2 and is thought to have overlapping functions. Mice lacking LAMP-1 had very minor defects compared to those deficient in LAMP-2 expression. However, the loss of both proteins resulted in embryonic lethality, suggesting that each protein possesses some unique and necessary functions.

Anti-LAMP-1 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 45 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: LAMP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR