Mouse Recombinant IL-2
Supplier: Rockland Immunochemical
010-001-V27-0100 010-001-V27-0020 010-001-V27-0005
75934-724EA 4283 USD
75934-724 75934-726 75934-728 75934-730
Mouse Recombinant IL-2
Proteins and Peptides
Recombinant Mouse IL-2 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin-2 (IL-2) is an immuno-modulatory cytokine produced by stimulated T cells. IL-2 signals through the IL-2R receptor to induce proliferation of activated T cells. IL-2 also stimulates proliferation and differentiation of B cells, natural killer cells, monocytes and macrophages. Recombinant mouse IL-2 is a non-glycosylated protein, containing 149 amino acids, with a molecular weight of 17.2 kDa.
Human Interleukin-2 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Human Interleukin-2 in immunological assays.