Human Recombinant IL-17E (IL-25)
Supplier: Rockland Immunochemical
009-001-V57-0005 009-001-V57-0100 009-001-V57-0025
75929-092EA 154.63 USD
75929-092 75934-694 75934-702 75934-710
Human Recombinant IL-17E (IL-25)
Proteins and Peptides
Recombinant Human IL-17E (IL-25) control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin-17E (IL-17E), also commonly called IL-25, is a pro-inflammatory cytokine member of a six-species family of proteins (IL-17A-17F). IL-17F stimulates secretion of IL-8 and induces activation of NF-kB by binding to the receptor IL-17RB. Recombinant human IL-17E is a non-glycosylated, disulfide-linked, homodimer, containing two 145 amino acid chains, with a total molecular weight of 35.5 kDa.
Interleukin-17E Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-17E in immunological assays.