Horse Plasma Non Sterile In Alsever's, Rockland Immunochemicals
Supplier: Rockland Immunochemical
D407-02-1000 D407-02-0100 D407-02-0050 D407-02-0500
75934-484EA 1074.03 USD
75934-484 75934-486 75934-488 75934-490
Horse Plasma Non Sterile In Alsever's, Rockland Immunochemicals
Blood
Non-Sterile Plasma in Anticoagulant
  • prepared by collection of whole blood into said anticoagulant
  • Red blood cells are removed through a process of centrifugation and filtration techniques - leaving non-sterile plasma behind
used as a component of bioassays, immunoassays or enzyme assays
provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health
also available as a sterile preparation for use as a cell culture supplement
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal