Anti-TNFSF15 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-401-H25
75934-268EA 635.22 USD
75934-268
Anti-TNFSF15 Rabbit Polyclonal Antibody
Antibodies
TL1A antibody detects human TL1A. Members in the TNF and its receptor superfamilies regulate immune responses and induce apoptosis. DR3 (also termed Wsl-1, Apo-3, TRAMP, and LARD) is preferentially expressed by T lymphocytes and upregulated during T cell activation. The ligand for DR3 was recently identified and designated TL1A. TL1A also binds decoy receptor DcR3/TR6, which is expressed in several lung and colon carcinomas and in some normal tissues. TL1A induces apoptosis and NF-kappaB activation in DR3 expressing cells, which is antagonized by DcR3. TL1A is upregulated by proinflammatory cytokines TNF and IL-1. TL1A is a longer variant of TL1 (also called VEGI). Anti-TL1A antibodies are ideal for investigators involved in apoptosis, NFkappaB, cytokines and growth factors research.

Anti-TL1A Antibody is suitable for use in E, and WB. Expect a band approximately ~28.0 kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: TNFSF15
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR